Learn More
Alkali Scientific Creatine Phosphate Disodium Salt Tetrahydrate, 1 Kg
SDP
Catalog No. 5000660346
Encompass_Preferred
Creatine Phosphate Disodium Salt Tetrahydrate is a high-purity compound used in energy metabolism studies. The disodium salt tetrahydrate form is highly soluble and ideal for use in biochemical assays, particularly in laboratories studying muscle metabolism and energy production. Creatine phosphate plays a crucial role in regenerating ATP, the primary energy source for cellular functions, especially in muscle cells. It is widely used in clinical chemistry, sports science, and metabolic research. This compound is typically used in laboratory preparations, clinical research, and the development of energy metabolism studies to assess creatine levels in biological systems.
Catalog No. 50-006-60346 Supplier Alkali Scientific Supplier No. CRE002CP
Creatine Phosphate Disodium Salt Tetrahydrate is a high-purity compound used in energy metabolism studies. The disodium salt tetrahydrate form is highly soluble and ideal for use in biochemical assays, particularly in laboratories studying muscle metabolism and energy production. Creatine phosphate plays a crucial role in regenerating ATP, the primary energy source for cellular functions, especially in muscle cells. It is widely used in clinical chemistry, sports science, and metabolic research. This compound is typically used in laboratory preparations, clinical research, and the development of energy metabolism studies to assess creatine levels in biological systems.