Learn More
Alkali Scientific Creatine Phosphate Disodium Salt Tetrahydrate, 100 G
SDP
Catalog No. 5000660780
Encompass_Preferred
Creatine Phosphate Disodium Salt Tetrahydrate is a high-purity compound used in energy metabolism studies. This 100g powder form is commonly used in laboratory experiments involving muscle contraction, energy storage, and metabolic regulation. Creatine phosphate is an important energy reservoir in muscle cells, and it plays a key role in rapid energy transfer during intense physical activities. Researchers utilize creatine phosphate in enzymatic assays, cellular studies, and biochemistry research focused on energy metabolism and muscle biochemistry. It is also used in pharmaceutical research for developing treatments related to muscular disorders and energy metabolism regulation.
Catalog No. 50-006-60780 Supplier Alkali Scientific Supplier No. CRE001CP
Creatine Phosphate Disodium Salt Tetrahydrate is a high-purity compound used in energy metabolism studies. This 100g powder form is commonly used in laboratory experiments involving muscle contraction, energy storage, and metabolic regulation. Creatine phosphate is an important energy reservoir in muscle cells, and it plays a key role in rapid energy transfer during intense physical activities. Researchers utilize creatine phosphate in enzymatic assays, cellular studies, and biochemistry research focused on energy metabolism and muscle biochemistry. It is also used in pharmaceutical research for developing treatments related to muscular disorders and energy metabolism regulation.