Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Creatine phosphokinase, rabbit muscle (CPK) | 9001-15-4 | 1 KU
SDP

Catalog No. 5000397432
Encompass_Preferred

Creatine phosphokinase, rabbit muscle (CPK) catalyzes the reversible reaction of creatine and ATP to form phosphocreatine and ADP. This enzyme is crucial for maintaining a constant ATP/ADP ratio during rapid energy turnover.

  • Catalyzes reversible reaction of creatine and ATP
  • Forms phosphocreatine and ADP
  • Key enzyme for maintaining constant ATP/ADP ratio
  • Appearance: solid
  • Color: white to off-white
  • Enzyme activity: ≥100 U/mg solid
  • Unit definition: amount of enzyme that transfers 1.0 μmole of phosphate from phosphocreatine to ADP per min at pH 7.4 at 30°C
  • Solubility: can be dissolved and stored at 1 mg/ml using 20 mM Tris (pH 8.0)
  • Avoids repeated freezing and thawing
  • Recommended to add 0.1% albumin to reaction buffer when diluting to avoid inactivation

Catalog No. 50-003-97432 Supplier Medchemexpress LLC Supplier No. HYP27991KU
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Creatine phosphokinase, rabbit muscle (CPK) catalyzes the reversible reaction of creatine and ATP to form phosphocreatine and ADP. This enzyme is crucial for maintaining a constant ATP/ADP ratio during rapid energy turnover.

  • Catalyzes reversible reaction of creatine and ATP
  • Forms phosphocreatine and ADP
  • Key enzyme for maintaining constant ATP/ADP ratio
  • Appearance: solid
  • Color: white to off-white
  • Enzyme activity: ≥100 U/mg solid
  • Unit definition: amount of enzyme that transfers 1.0 μmole of phosphate from phosphocreatine to ADP per min at pH 7.4 at 30°C
  • Solubility: can be dissolved and stored at 1 mg/ml using 20 mM Tris (pH 8.0)
  • Avoids repeated freezing and thawing
  • Recommended to add 0.1% albumin to reaction buffer when diluting to avoid inactivation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.