Learn More
Medchemexpress LLC Creatine (Standard) | 57-00-1 | 100.0% | 131.13 | 25 MG
SDP
Catalog No. 5000453245
Encompass_Preferred
Creatine (Standard) is the analytical standard of Creatine. Creatine is an endogenous amino acid derivative that plays an important role in cellular energy, especially in muscle and brain.
  • Used as an analytical standard
  • Suitable for qualitative research experiments
  • Suitable for quantitative research experiments
  • Suitable for methodological research experiments in HPLC, GC, and MS
Catalog No. 50-004-53245 Supplier Medchemexpress LLC Supplier No. 25MGHYW010388R
Creatine (Standard) is the analytical standard of Creatine. Creatine is an endogenous amino acid derivative that plays an important role in cellular energy, especially in muscle and brain.
  • Used as an analytical standard
  • Suitable for qualitative research experiments
  • Suitable for quantitative research experiments
  • Suitable for methodological research experiments in HPLC, GC, and MS