Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Creatine (standard) | 57-00-1 | 99.98% | 131.13 | 250 MG
SDP

Catalog No. 5000451371
Encompass_Preferred

Creatine (Standard) is the analytical standard of Creatine, intended for research and analytical applications. Creatine is an endogenous amino acid derivative that plays an important role in cellular energy, particularly in muscle and brain.

  • Analytical standard for Creatine.
  • Intended for research and analytical applications.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Endogenous metabolite.
  • Formula: C4H9N3O2.
  • Appearance: Solid, white to off-white.
  • Structure classification: Amino acids.
  • Initial source: Endogenous metabolite, Disease markers (Cancer, Nervous System Disorder), Human Gut Microbiota Metabolites.

Catalog No. 50-004-51371 Supplier Medchemexpress LLC Supplier No. 250MGHYW010388R
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Creatine (Standard) is the analytical standard of Creatine, intended for research and analytical applications. Creatine is an endogenous amino acid derivative that plays an important role in cellular energy, particularly in muscle and brain.

  • Analytical standard for Creatine.
  • Intended for research and analytical applications.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Endogenous metabolite.
  • Formula: C4H9N3O2.
  • Appearance: Solid, white to off-white.
  • Structure classification: Amino acids.
  • Initial source: Endogenous metabolite, Disease markers (Cancer, Nervous System Disorder), Human Gut Microbiota Metabolites.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.