Learn More
Medchemexpress LLC Creatine (Standard) | 57-00-1 | 99.98% | 131.13 | 50 MG
SDP
Catalog No. 5000452561
Encompass_Preferred
Creatine (Standard) is an analytical standard of Creatine, an endogenous amino acid derivative crucial for cellular energy in muscle and brain. It is designed for research and analytical applications, serving as a reference in various assays.
  • Used in qualitative research experiments.
  • Used in quantitative research experiments.
  • Used in methodological research experiments.
  • Applicable in HPLC.
  • Applicable in GC.
  • Applicable in MS.
Catalog No. 50-004-52561 Supplier Medchemexpress LLC Supplier No. HYW010388R50MG
Creatine (Standard) is an analytical standard of Creatine, an endogenous amino acid derivative crucial for cellular energy in muscle and brain. It is designed for research and analytical applications, serving as a reference in various assays.
  • Used in qualitative research experiments.
  • Used in quantitative research experiments.
  • Used in methodological research experiments.
  • Applicable in HPLC.
  • Applicable in GC.
  • Applicable in MS.