Learn More
Cayman Chemical Creatine (hydrate), Purity: ≥95%, CAS: 6020-87-7, 100 g
SDP
Catalog No. 502915196
Encompass_Preferred
A derivative of glycine; phosphorylated to creatine phosphate, which is used as an energy reservoir; decreased brain levels are associated with inborn errors of creatine metabolism characterized by deficiency of AGAT, GAMT, or the creatine transporter
Catalog No. 50-291-5196 Supplier Cayman Chemical Supplier No. 36940100 G
A derivative of glycine; phosphorylated to creatine phosphate, which is used as an energy reservoir; decreased brain levels are associated with inborn errors of creatine metabolism characterized by deficiency of AGAT, GAMT, or the creatine transporter