Learn More
Medchemexpress LLC Creatinine-13C | 1173022-95-1 | 99.9% | 1 MG
SDP
Catalog No. 5000408033
Encompass_Preferred
Creatinine-13C is a 13C-labeled version of Creatinine. Creatinine, also known as NSC13123, is a break-down product of creatine phosphate found in muscle. It is typically produced by the body at a fairly constant rate.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Isotope-labeled compound
Catalog No. 50-004-08033 Supplier Medchemexpress LLC Supplier No. HYB0504S11MG
Creatinine-13C is a 13C-labeled version of Creatinine. Creatinine, also known as NSC13123, is a break-down product of creatine phosphate found in muscle. It is typically produced by the body at a fairly constant rate.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Isotope-labeled compound