Learn More
Medchemexpress LLC Creatinine-13C | 1173022-95-1 | >99.9% | 5 MG
SDP
Catalog No. 5000406737
Encompass_Preferred
Creatinine-13C is the 13C-labeled Creatinine, a breakdown product of creatine phosphate in muscle that is usually produced at a fairly constant rate by the body. This product is intended for research use only and serves as a valuable tool in various analytical and research applications due to its stable heavy isotope label. It is particularly useful for precise quantitative analysis in studies involving pharmacokinetics and metabolic profiling.
  • Used as a tracer in research studies.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes of carbon for drug development processes.
  • Can influence pharmacokinetic and metabolic profiles of drugs.
  • Molecular formula is C313CH7N3O.
Catalog No. 50-004-06737 Supplier Medchemexpress LLC Supplier No. HYB0504S15MG
Creatinine-13C is the 13C-labeled Creatinine, a breakdown product of creatine phosphate in muscle that is usually produced at a fairly constant rate by the body. This product is intended for research use only and serves as a valuable tool in various analytical and research applications due to its stable heavy isotope label. It is particularly useful for precise quantitative analysis in studies involving pharmacokinetics and metabolic profiling.
  • Used as a tracer in research studies.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes of carbon for drug development processes.
  • Can influence pharmacokinetic and metabolic profiles of drugs.
  • Molecular formula is C313CH7N3O.