Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Organic Chemistry Creatinine | 1kg | 60-27-5 | MFCD00059730

Catalog No. C42551KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass

Creatinine , 1kg

About this Item:

CAS #: 60-27-5
MDL #: MFCD00059730
Molecular Weight: 113.12
UNSPSC Code: 12352100
Chemical Formula: C4H7N3O

Catalog No. C42551KG Supplier Sigma Organic Chemistry Supplier No. C42551KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Creatinine , 1kg

About this Item:

CAS #: 60-27-5
MDL #: MFCD00059730
Molecular Weight: 113.12
UNSPSC Code: 12352100
Chemical Formula: C4H7N3O

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.