Learn More
MOLECULAR DEPOT LLC Creatinine - 2/1 mL
Catalog No. 5000013464
Encompass_Preferred
Creatinine for Interference Testing consists of a highly concentrated solution of Creatinine ( 50 mg/mL) provided with the corresponding diluent (diluent A) This solution can be spiked into specimens to test Creatinine interference/tolerance in assays For in vitro research use only
Catalog No. 50-000-13464 Supplier MOLECULAR DEPOT LLC Supplier No. K2010008C
Creatinine for Interference Testing consists of a highly concentrated solution of Creatinine ( 50 mg/mL) provided with the corresponding diluent (diluent A) This solution can be spiked into specimens to test Creatinine interference/tolerance in assays For in vitro research use only