Learn More
Enzo Life Sciences Creatinine colorimetric detection kit (2x96wells)
SDP
Catalog No. NC0203788
Encompass_Preferred
Low price, rapid creatinine detection kit for diabetes, cardiovascular and renal research. The Creatinine colorimetric detection kit utilizes a single-step liquid detection reagent that is safer and less time consuming than other assay methods. This kit is calibrated against the NIST standard and offers reproducible results with less than 6% inter- and intra-assay variation. Sensitivity: 0.042 mg/dl (range 0.31 - 20 mg/dl). Assay Time: 30 minutes. Applications: Colorimetric detection. Species Reactivity: Species independent. Contents: Microtiter plate, Standard, Detection Reagent
Catalog No. NC0203788 Supplier Enzo Life Sciences Supplier No. ADI907030A
Low price, rapid creatinine detection kit for diabetes, cardiovascular and renal research. The Creatinine colorimetric detection kit utilizes a single-step liquid detection reagent that is safer and less time consuming than other assay methods. This kit is calibrated against the NIST standard and offers reproducible results with less than 6% inter- and intra-assay variation. Sensitivity: 0.042 mg/dl (range 0.31 - 20 mg/dl). Assay Time: 30 minutes. Applications: Colorimetric detection. Species Reactivity: Species independent. Contents: Microtiter plate, Standard, Detection Reagent