Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Creatinine

Catalog No. 502907781 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 mg
10 g
25 g
100 g
1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-290-7781 1 kg
C4255100G 100 g
C425510G 10 g
C425510MG 10 mg
C425525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 50-290-7781 Supplier Sigma Aldrich Supplier No. C42551KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Creatinine
TRUSTED_SUSTAINABILITY

Specifications

CAS 60-27-5
Melting Point 295°C (lit.)
Molecular Formula C4H7N3O
Linear Formula C4H7N3O
MDL Number MFCD00059730
Quantity 1 kg
Synonym 2-Amino-1-methyl-2-imidazolin-4-one; 2-Imino-1-methylimidazolidin-4-one; 2-Imino-N-methylhydantoin
Molecular Weight (g/mol) 113.12
Percent Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.