Learn More
Medchemexpress LLC Creatinine | 60-27-5 | 99.9% | 1 ML
SDP
Catalog No. 5000405535
Encompass_Preferred
Creatinine is an orally active, blood-brain barrier-permeable modulator of GABAA and NMDA receptors, exhibiting antioxidation and metabolic regulation activities. Generated from creatine and phosphocreatine, its serum levels serve as a crucial biomarker for renal function, muscle mass, and clinical outcomes. This compound is widely utilized in research concerning neurodegenerative diseases, psychiatric disorders, acute kidney injury, chronic kidney disease, and renal failure.
  • Orally active and blood-brain barrier-permeable
  • Modulates GABAA and NMDA receptors
  • Exhibits antioxidation and metabolic regulation activities
  • Crucial biomarker for renal function, muscle mass, and clinical outcomes
  • Applied in research for neurodegenerative diseases, psychiatric disorders, and various kidney conditions
Catalog No. 50-004-05535 Supplier Medchemexpress LLC Supplier No. HYB050410MM/1ML
Creatinine is an orally active, blood-brain barrier-permeable modulator of GABAA and NMDA receptors, exhibiting antioxidation and metabolic regulation activities. Generated from creatine and phosphocreatine, its serum levels serve as a crucial biomarker for renal function, muscle mass, and clinical outcomes. This compound is widely utilized in research concerning neurodegenerative diseases, psychiatric disorders, acute kidney injury, chronic kidney disease, and renal failure.
  • Orally active and blood-brain barrier-permeable
  • Modulates GABAA and NMDA receptors
  • Exhibits antioxidation and metabolic regulation activities
  • Crucial biomarker for renal function, muscle mass, and clinical outcomes
  • Applied in research for neurodegenerative diseases, psychiatric disorders, and various kidney conditions