Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Creatinine | 60-27-5 | 99.99% | 113.12 | C4H7N3O | 10 MG
SDP

Catalog No. 5000297364
Encompass_Preferred

Creatinine (standard) is an analytical reference material supplied as a high-purity solid for research and analytical laboratories. It is used for calibration, method validation, and quality control in quantitative and qualitative creatinine analysis.

  • High purity (99.99%) suitable for analytical work.
  • CAS number 60-27-5.
  • Molecular weight 113.12 g/mol.
  • Supplied as a solid in small sizes down to 10 MG.
  • Intended for calibration, method validation, and quality control.
  • COA and SDS available for traceability and safety.

Catalog No. 50-002-97364 Supplier Medchemexpress LLC Supplier No. HYB0504R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Creatinine (standard) is an analytical reference material supplied as a high-purity solid for research and analytical laboratories. It is used for calibration, method validation, and quality control in quantitative and qualitative creatinine analysis.

  • High purity (99.99%) suitable for analytical work.
  • CAS number 60-27-5.
  • Molecular weight 113.12 g/mol.
  • Supplied as a solid in small sizes down to 10 MG.
  • Intended for calibration, method validation, and quality control.
  • COA and SDS available for traceability and safety.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.