Learn More
Medchemexpress LLC Creatinine | 60-27-5 | MFCD00004277 | 99.99% | 25 MG
SDP
Catalog No. 5000450484
Encompass_Preferred
Creatinine (Standard) is an analytical standard of Creatinine, a breakdown product of creatine phosphate found in muscles. It is commonly used for research and analytical applications, particularly in qualitative, quantitative, and methodological research experiments involving techniques such as HPLC, GC, and MS.
  • Analytical standard for Creatinine
  • Suitable for research and analytical applications
  • Utilized in HPLC, GC and MS experiments
  • Derived from creatine phosphate breakdown
  • Store at 2-8°C, protected from light.
Catalog No. 50-004-50484 Supplier Medchemexpress LLC Supplier No. 25MGHYB0504R
Creatinine (Standard) is an analytical standard of Creatinine, a breakdown product of creatine phosphate found in muscles. It is commonly used for research and analytical applications, particularly in qualitative, quantitative, and methodological research experiments involving techniques such as HPLC, GC, and MS.
  • Analytical standard for Creatinine
  • Suitable for research and analytical applications
  • Utilized in HPLC, GC and MS experiments
  • Derived from creatine phosphate breakdown
  • Store at 2-8°C, protected from light.