Learn More
MOLECULAR DEPOT LLC Creatinine BSA - 0.5 mg
Catalog No. 5000013349
Encompass_Preferred
Creatinine BSA Conjugate Solution (0 5 mg) is a concentrated solution of Bovine Serum Albumin (BSA) conjugated to Creatinine (a breakdown product of creatine phosphate) A carbon spacer arm links BSA to the Creatinine moiety Each BSA molecule contains several Creatinine units Dilutions of the provided conjugate can be made in Phosphate Buffer Saline pH 7 4
Catalog No. 50-000-13349 Supplier MOLECULAR DEPOT LLC Supplier No. P2010017
Creatinine BSA Conjugate Solution (0 5 mg) is a concentrated solution of Bovine Serum Albumin (BSA) conjugated to Creatinine (a breakdown product of creatine phosphate) A carbon spacer arm links BSA to the Creatinine moiety Each BSA molecule contains several Creatinine units Dilutions of the provided conjugate can be made in Phosphate Buffer Saline pH 7 4