Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diazyme Laboratories Creatinine Calibrator

Catalog No. 23700564
Encompass
Encompass_Preferred
Change view
Click to view available options
Format:
1 x 1 mL Vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
23-700-564 1 x 1 mL Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-700-564 Supplier Diazyme Laboratories Supplier No. DZ072BCAL
May include imposed supplier surcharges.
Add to Cart
Add to Cart
This item is not returnable. View return policy

Diazyme Creatinine Calibrator Set is used for the testing of Creatinine in Serum and Urine.

  • Offers significant advantages compared to the conventional Jaffe method for Creatinine, which is caustic (picric acid)  and can stain analyzer tubing and cuvettes
  • Enzymatic method is non-caustic, non-staining and has no costly hazardous shipping requirements
  • Unlike the Jaffe method Diazyme's enzymatic assay shows that the following substances normally present in serum produced less than 10% deviation at the listed concentrations: Triglyceride at 1000 mg/dL, Ascorbic Acid at 10 mM, Bilirubin at 40 mg/dL, Bilirubin Conjugate at 30 mg/dL, Hemoglobin at 500 mg/dL
  • The following substances normally present in urine produced less than 10% deviation at the listed concentrations: Triglycerides at 1000 mg/dL, Ascorbic Acid at 10 mM, Bilirubin at 40 mg/dL, Bilirubin Conjugate at 40 mg/dL, Hemoglobin at 1000 mg/dL
  • Packaged for optimal operator convenience, reagent transfer can be eliminated for most chemistry systems with instrument specific packaging options including Roche™ Hitachi 917 series, Beckman AU (400/600/640/680), Beckman Synchron CX

Specifications

Certifications/Compliance 510(k) Cleared, EU: CE IVD
Content And Storage Store at 2°C to 8°C. DO NOT FREEZE. Note: Store CALIBRATOR tightly capped when not in use.
Description Creatinine Assay Calibrator Set
Concentration Lab dependent
Format 1 x 1 mL Vial
Type Diagnostic
Includes Supplied in liquid stable form (1 x 1 mL). The CALIBRATOR is manufactured from a buffer base. The concentration of the calibrator is lot specific and the creatinine concentration are expressed in μmol.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.