Learn More
Medchemexpress LLC Creatinine-d3 | 143827-20-7 | MFCD00190410 | 99.9% | 116.14 | C4H4D3N3O | 10 MG
SDP
Catalog No. 5000298290
Encompass_Preferred
Creatinine-d3 is a deuterium-labeled form of creatinine used as a tracer and internal standard for quantitative analysis by NMR, GC-MS, and LC-MS. It has molecular formula C4H4D3N3O and a molecular weight of 116.14 g/mol. For research use only.
  • Deuterated internal standard for accurate quantitation.
  • High isotopic and chemical purity (99.9% by LCMS) for reliable results.
  • Compatible with NMR, GC-MS, and LC-MS analytical methods.
  • Available in small research pack sizes to support analytical workflows.
Catalog No. 50-002-98290 Supplier Medchemexpress LLC Supplier No. HYB0504S10MG
Creatinine-d3 is a deuterium-labeled form of creatinine used as a tracer and internal standard for quantitative analysis by NMR, GC-MS, and LC-MS. It has molecular formula C4H4D3N3O and a molecular weight of 116.14 g/mol. For research use only.
  • Deuterated internal standard for accurate quantitation.
  • High isotopic and chemical purity (99.9% by LCMS) for reliable results.
  • Compatible with NMR, GC-MS, and LC-MS analytical methods.
  • Available in small research pack sizes to support analytical workflows.