Learn More
Medchemexpress LLC Creatinine-d5 | 60-27-5 | 99.9% | 1 MG
SDP
Catalog No. 5000408034
Encompass_Preferred
Creatinine-d5 is the deuterium-labeled version of Creatinine. Creatinine is a breakdown product of creatine phosphate found in muscle, and the body typically produces it at a consistent rate. This compound is intended for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis through methods like NMR, GC-MS, or LC-MS
  • Stable heavy isotopes can be incorporated into drug molecules for use as tracers in drug development for quantitation
  • Deuteration is being explored for its potential to impact the pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-08034 Supplier Medchemexpress LLC Supplier No. HYB0504S21MG
Creatinine-d5 is the deuterium-labeled version of Creatinine. Creatinine is a breakdown product of creatine phosphate found in muscle, and the body typically produces it at a consistent rate. This compound is intended for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis through methods like NMR, GC-MS, or LC-MS
  • Stable heavy isotopes can be incorporated into drug molecules for use as tracers in drug development for quantitation
  • Deuteration is being explored for its potential to impact the pharmacokinetic and metabolic profiles of drugs