Learn More
Medchemexpress LLC Creatinine-d5 | 60-27-5 | 99.9% | 5 MG
SDP
Catalog No. 5000405277
Encompass_Preferred
Creatinine-d5 is the deuterium labeled version of Creatinine, a breakdown product of creatine phosphate found in muscle. It is typically produced at a consistent rate by the body. Intended for research use only, this compound serves as a tracer and internal standard in analytical techniques.
  • Deuterium labeled version of creatinine
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Suitable for techniques like NMR, GC-MS, or LC-MS
  • May influence pharmacokinetic and metabolic profiles of drugs
  • For research use only
Catalog No. 50-004-05277 Supplier Medchemexpress LLC Supplier No. HYB0504S25MG
Creatinine-d5 is the deuterium labeled version of Creatinine, a breakdown product of creatine phosphate found in muscle. It is typically produced at a consistent rate by the body. Intended for research use only, this compound serves as a tracer and internal standard in analytical techniques.
  • Deuterium labeled version of creatinine
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Suitable for techniques like NMR, GC-MS, or LC-MS
  • May influence pharmacokinetic and metabolic profiles of drugs
  • For research use only