Learn More
Medchemexpress LLC Creatinine-d5 | 60-27-5 | MFCD28009754 | 99.9% | 118.15 g/mol | C4H2D5N3O | 10 MG
SDP
Catalog No. 5000298101
Encompass_Preferred
Creatinine-d5 is a deuterium-labeled internal standard of creatinine used as an analytical reference for isotope-dilution mass spectrometry and other biochemical assays. It provides a stable, isotopically enriched analogue for accurate quantification and method validation in research applications.
  • Deuterium-labeled internal standard for LC-MS/MS quantification.
  • High isotopic enrichment suitable for analytical accuracy.
  • Supplied as a small, stable solid for easy handling and dissolution.
  • Intended for research use only; not for human or diagnostic use.
Catalog No. 50-002-98101 Supplier Medchemexpress LLC Supplier No. HYB0504S210MG
Creatinine-d5 is a deuterium-labeled internal standard of creatinine used as an analytical reference for isotope-dilution mass spectrometry and other biochemical assays. It provides a stable, isotopically enriched analogue for accurate quantification and method validation in research applications.
  • Deuterium-labeled internal standard for LC-MS/MS quantification.
  • High isotopic enrichment suitable for analytical accuracy.
  • Supplied as a small, stable solid for easy handling and dissolution.
  • Intended for research use only; not for human or diagnostic use.