Learn More
DIASYS DIAGNOSTIC SYSTEMS USA LLC DiaSys Creatinine Jaffe FS is a ready-to-use two component photometric reagent to determine Creatinine in serum, heparin plasma or urine on photometric systems. Each set contains R1=800mL, R2=200mL
Catalog No. NC2874315
Encompass_Preferred
Diagnostic reagent for quantitative in vitro determination of creatinine in animal serum, heparin plasma or urine on photometric systems. For veterinary or research use only. Creatinine values are determined to estimate the glomerular filtration rate, an indicator of kidney status. Method: DiaSys Creatinine Jaffe FS is based on the kinetic Jaffé method without deproteinization. The reagent is a liquid-stable kinetic test with a wide measuring range and freedom from lipemia and hemoglobin interference. Reagent 1 contains TRIS, Mg2+, ATP and NAD, reagent 2 contains Mg2+, Hexokinase and Glucose-6-Phospatedehydrogenase. Reagents must be stored refrigerated.
Catalog No. NC2874315 Supplier DIASYS DIAGNOSTIC SYSTEMS USA LLC Supplier No. 117119911887
Diagnostic reagent for quantitative in vitro determination of creatinine in animal serum, heparin plasma or urine on photometric systems. For veterinary or research use only. Creatinine values are determined to estimate the glomerular filtration rate, an indicator of kidney status. Method: DiaSys Creatinine Jaffe FS is based on the kinetic Jaffé method without deproteinization. The reagent is a liquid-stable kinetic test with a wide measuring range and freedom from lipemia and hemoglobin interference. Reagent 1 contains TRIS, Mg2+, ATP and NAD, reagent 2 contains Mg2+, Hexokinase and Glucose-6-Phospatedehydrogenase. Reagents must be stored refrigerated.