Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diazyme Laboratories Creatinine Liquid Reagent Assay

Catalog No. 23700646
Encompass
Change view
Click to view available options
Creatinine Liquid Reagent Assay
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
23-700-646 Creatinine Liquid Reagent Assay
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-700-646 Supplier Diazyme Laboratories Supplier No. DZ072BKY1
May include imposed supplier surcharges.
Add to Cart
Add to Cart
This item is not returnable. View return policy

Liquid, stable

  • Non-caustic, non-staining method
  • For quantitative determination of creatinine in serum and urine
  • Used in treatment of renal disease, monitoring renal dialysis and calculation basis for measuring other urine analytes

Specifications

Certifications/Compliance 510(k) cleared, EU: CE
Type Creatinine Liquid Reagent Assay
For Use With (Application) For treatment with renal disease, monitoring renal dialysis and calculation basis for measuring other urine analytes
Includes R1: 2 x 60 mL, R2: 2 x 20 mL
Form Liquid

For in vitro diagnostic use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.