Learn More
Sigma Aldrich Fine Chemicals Biosciences Creatinine NISTR SRMR 914b
Catalog No. 502908002
Encompass
This Standard Reference Material (SRM) is certified as a chemical of known purity. It is intended primarily for use in the calibration and standardization of procedures used for determination of creatinine concentration and for routine evaluations of daily working standards used in these procedures. For more information please refer to the SDS and COA. SRM 914B SDSSRM 914B cert
Catalog No. 50-290-8002 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. NIST914B
This Standard Reference Material (SRM) is certified as a chemical of known purity. It is intended primarily for use in the calibration and standardization of procedures used for determination of creatinine concentration and for routine evaluations of daily working standards used in these procedures. For more information please refer to the SDS and COA. SRM 914B SDSSRM 914B cert