Learn More
Cayman Chemical Creatinine Sodium Hydroxide, 5 ea
SDP
Catalog No. 502886674
Encompass_Preferred
Creatinine sodium hydroxide has been tested and formulated to work exclusively with Cayman’s Creatinine (urinary) Colorimetric Assay Kit (Item No. 500701).
Catalog No. 50-288-6674 Supplier Cayman Chemical Supplier No. 100053165 EA
Creatinine sodium hydroxide has been tested and formulated to work exclusively with Cayman’s Creatinine (urinary) Colorimetric Assay Kit (Item No. 500701).