Learn More
Medchemexpress LLC Creatinine (Standard) | 60-27-5 | 99.99% | 50 MG
SDP
Catalog No. 5000405538
Encompass_Preferred
Creatinine (Standard) is an analytical standard intended for research and analytical applications. As a breakdown product of creatine phosphate in muscles, it serves as a reference standard for various assays.
  • Analytical standard of creatinine
  • Commonly used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications
Catalog No. 50-004-05538 Supplier Medchemexpress LLC Supplier No. HYB0504R50MG
Creatinine (Standard) is an analytical standard intended for research and analytical applications. As a breakdown product of creatine phosphate in muscles, it serves as a reference standard for various assays.
  • Analytical standard of creatinine
  • Commonly used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications