Learn More
Enzo Life Sciences CREB binding protein (catalytic domain) (human), (recombinant) (100 µg)
SDP
Supplier:  Enzo Life Sciences BMLSE4520100
Encompass_Preferred
Alternative name: CBP. MW: 45.4 kDa. Formulation: Liquid. In 50mM TRIS-HCl, pH 8.0, containing 0.1mM EDTA, 10% glycerol. Source: Produced in E. coli. Catalytic domain (aa 1319-1710) of human CBP. UniProt: Q92793. Handling: Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -80°C. Long Term Storage: -80°C
Catalog No. NC0738768