Learn More
Aviva Systems Biology Corp CREB1 Antibody - N-terminal region (ARP31264_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31264_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CREB1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CREB1. Alternate names: CREB, MGC9284ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54191