Learn More
Aviva Systems Biology Corp CREB3L1 Antibody - N-terminal region (ARP36109_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36109_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CREB3L1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CREB3L1. Alternate names: OASISApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-53029