Learn More
Aviva Systems Biology Corp CREBL1 Antibody - middle region (ARP31562_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31562_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CREBL1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CREBL1. Alternate names: CREB-RP, FLJ10066, G13, CREBL1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-54267