Learn More
CPC Scientific H-Lys-Arg-Arg-Glu-Ile-Leu-Ser-Arg-Arg-Pro-Ser-Tyr-Arg-OH (trifluoroacetate salt) 1MG
SDP
Catalog No. 502110487
Encompass
SEQUENCE: H-Lys-Arg-Arg-Glu-Ile-Leu-Ser-Arg-Arg-Pro-Ser-Tyr-Arg-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: KRREILSRRPSYRMOLECULAR FORMULA: C73H129N29O19MOLECULAR WEIGHT: 1716.99STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [149155-45-3]SYNONYMS: RESEARCH AREA: ApoptosisREFERENCES:
Colbran, Janet L., et al. Biochemistry and Cell Biology 70.10-11 (1992): 1277-1282.SKU(s): SUBS-013A, SUBS-013B
Catalog No. 50-211-0487 Supplier CPC Scientific Supplier No. SUBS013B
SEQUENCE: H-Lys-Arg-Arg-Glu-Ile-Leu-Ser-Arg-Arg-Pro-Ser-Tyr-Arg-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: KRREILSRRPSYRMOLECULAR FORMULA: C73H129N29O19MOLECULAR WEIGHT: 1716.99STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [149155-45-3]SYNONYMS: RESEARCH AREA: ApoptosisREFERENCES:
Colbran, Janet L., et al. Biochemistry and Cell Biology 70.10-11 (1992): 1277-1282.SKU(s): SUBS-013A, SUBS-013B