Learn More
Enzo Life Sciences CREBtide (1mg)
SDP
Supplier:  Enzo Life Sciences BMLP1950001
Encompass_Preferred
This peptide, KRREILSRRPSYR, is an excellent substrate for many serine kinases, including PKA and PKC. Purity: ≥95% (HPLC). Formulation: Lyophilized. Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-201-1273