Learn More
Medchemexpress LLC CREBtide TFA | 98.7% | 1716.99 (free acid) | 5 MG
SDP
Catalog No. 5000424018
Encompass_Preferred
CREBtide TFA is a synthetic 13 amino acid peptide, a CREB (cAMP response element binding protein)-like peptide. It functions as a substrate for both Protein Kinase A (PKA) and Protein Kinase C (PKC).
  • Solid appearance
  • White to off-white color
  • Amino acid sequence: Lys-Arg-Arg-Glu-Ile-Leu-Ser-Arg-Arg-Pro-Ser-Tyr-Arg
  • Reported as a PKA and PKC substrate
  • Apparent Km of 3.9 μM for phosphorylation by cAK
  • Apparent Km of 2.9 μM for phosphorylation by cGK
Catalog No. 50-004-24018 Supplier Medchemexpress LLC Supplier No. HYP1595A5MG
CREBtide TFA is a synthetic 13 amino acid peptide, a CREB (cAMP response element binding protein)-like peptide. It functions as a substrate for both Protein Kinase A (PKA) and Protein Kinase C (PKC).
  • Solid appearance
  • White to off-white color
  • Amino acid sequence: Lys-Arg-Arg-Glu-Ile-Leu-Ser-Arg-Arg-Pro-Ser-Tyr-Arg
  • Reported as a PKA and PKC substrate
  • Apparent Km of 3.9 μM for phosphorylation by cAK
  • Apparent Km of 2.9 μM for phosphorylation by cGK