Learn More
PELICAN BIOTHERMAL LLC CREDO CUBE 12L
Supplier:  PELICAN BIOTHERMAL LLC CR12A20M1296WTU
Encompass_Preferred
SERIES 20M, SINGLE VIP, WHITE PLASTIC OUTER.Temperature Range Sub -18°C.Duration 96.00 hours
Catalog No. NC1876376