Learn More
Medchemexpress LLC CREKA peptide acetate | 665.76 | 50 MG
SDP
Catalog No. 5000425210
Encompass_Preferred
CREKA peptide acetate is a short peptide sequence that belongs to self-assembling peptides (SAPs). It can self-assemble into functional nanostructures, such as nanofibers, under physiological conditions. This peptide acetate can be used to target tumor cells and tumor vasculature, demonstrating antitumor activity.
  • Short peptide sequence
  • Belongs to self-assembling peptides (SAPs)
  • Can self-assemble into functional nanostructures (nanofibers) under physiological conditions
  • Can be used to target tumor cells and tumor vasculature
  • Exhibits antitumor activity
  • For research use only
Catalog No. 50-004-25210 Supplier Medchemexpress LLC Supplier No. HYP10709A50MG
CREKA peptide acetate is a short peptide sequence that belongs to self-assembling peptides (SAPs). It can self-assemble into functional nanostructures, such as nanofibers, under physiological conditions. This peptide acetate can be used to target tumor cells and tumor vasculature, demonstrating antitumor activity.
  • Short peptide sequence
  • Belongs to self-assembling peptides (SAPs)
  • Can self-assemble into functional nanostructures (nanofibers) under physiological conditions
  • Can be used to target tumor cells and tumor vasculature
  • Exhibits antitumor activity
  • For research use only