Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CREKA peptide acetate | 847058-45-1 | 98.1% | 605.71 | 5 MG
SDP

Catalog No. 5000424951
Encompass_Preferred

CREKA peptide acetate | 847058-45-1 | 98.1% | 605.71 | 5 MG

Catalog No. 50-004-24951 Supplier Medchemexpress LLC Supplier No. HYP10709A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CREKA peptide acetate | 847058-45-1 | 98.1% | 605.71 | 5 MG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.