Learn More
Aviva Systems Biology Corp CRELD1 Antibody - C-terminal region (ARP45004_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45004_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CRELD1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human CRELD1. Alternate names: AVSD2, CIRRIN, DKFZP566D213ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-53441