Learn More
ABclonal Technology CRELD1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A15186500UL
Encompass_Preferred
This gene encodes a member of a subfamily of epidermal growth factor-related proteins. The encoded protein is characterized by a cysteine-rich with epidermal growth factor-like domain. This protein may function as a cell adhesion molecule. Mutations in this gene are the cause of atrioventricular septal defect. Alternate splicing results in multiple transcript variants.
Catalog No. 50-001-05500