Learn More
Aviva Systems Biology Corp CRELD2 Antibody - middle region (ARP62266_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62266_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CRELD2. It was validated on Western Blot. . Alternate names: DKFZp667O055, MGC11256ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-287-637