Learn More
Selleck Chemical LLC Cremophor EL-50g
SDP
Supplier:  Selleck Chemical LLC S6828
Encompass_Preferred
Cremophor EL (CrEL, Kolliphor EL), a polyethoxylated surfactant, is a formulation vehicle used for various poorly-water soluble drugs, including the anticancer agent paclitaxel (Taxol). Cremophor EL is used in severe anaphylactoid hypersensitivity reactions, hyperlipidaemia, abnormal lipoprotein patterns, aggregation of erythrocytes and peripheral neuropathy.
Catalog No. 50-238-9899