Learn More
Cayman Chemical CrenIgacestat 10mg
SDP
Catalog No. 502938193
Encompass_Preferred
A Notch inhibitor; induces cell cycle arrest at the G0/G1 phase and decreases MYC and CCNA1 expression in Caki human renal cancer cells; reduces tumor growth and increases survival in a 769-P mouse xenograft model at 8 mg/kg per day
Catalog No. 50-293-8193 Supplier Cayman Chemical Supplier No. 2791910 MG
A Notch inhibitor; induces cell cycle arrest at the G0/G1 phase and decreases MYC and CCNA1 expression in Caki human renal cancer cells; reduces tumor growth and increases survival in a 769-P mouse xenograft model at 8 mg/kg per day