Learn More
Medchemexpress LLC HY-13223 10mg Medchemexpress, Crenolanib CAS:670220-88-9 Purity:>98%
SDP
Catalog No. 502028961
Encompass_Preferred
Medchemexpress, HY-13223 10mg Crenolanib CAS:670220-88-9 Crenolanib is a potent and selective inhibitor of wild-type and mutant isoforms of the class III receptor tyrosine kinases FLT3 and PDGFRα/β with Kds of 0.74 nM and 2.1 nM/3.2 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8961 Supplier Medchemexpress LLC Supplier No. HY1322310MG
Add to Cart
Add to Cart
Medchemexpress, HY-13223 10mg Crenolanib CAS:670220-88-9 Crenolanib is a potent and selective inhibitor of wild-type and mutant isoforms of the class III receptor tyrosine kinases FLT3 and PDGFRα/β with Kds of 0.74 nM and 2.1 nM/3.2 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.