Learn More
Medchemexpress LLC HY-13223 100mg , Crenolanib CP-868596 CAS:670220-88-9 Purity:98%
SDP
Catalog No. 501492867
Encompass_Preferred
Medchemexpress, HY-13223 100mg Crenolanib CP-868596 CAS:670220-88-9 Formula:C26H29N5O2 Kd: 2.1 nM (PDGFRα), 3.2 nM (PDGFRβ), 0.74 nM (FLT3) Purity:98% Crenolanib is a potent and selective inhibitor of PDGFRα/β , FLT3 with K d of 2.1 nM/3.2 nM, 0.74 nM, respectively, sensitive to D842V mutation not V561D mutation, and > 100-fold more selective for PDGFR than c-Kit, VEGFR-2, TIE-2, FGFR-2, EGFR, erbB2, and Src. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2867 Supplier Medchemexpress LLC Supplier No. HY13223100MG
Medchemexpress, HY-13223 100mg Crenolanib CP-868596 CAS:670220-88-9 Formula:C26H29N5O2 Kd: 2.1 nM (PDGFRα), 3.2 nM (PDGFRβ), 0.74 nM (FLT3) Purity:98% Crenolanib is a potent and selective inhibitor of PDGFRα/β , FLT3 with K d of 2.1 nM/3.2 nM, 0.74 nM, respectively, sensitive to D842V mutation not V561D mutation, and > 100-fold more selective for PDGFR than c-Kit, VEGFR-2, TIE-2, FGFR-2, EGFR, erbB2, and Src. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.