Learn More
Selleck Chemical LLC Crenolanib S2730-50mg
SDP
Catalog No. 503112232
Encompass_Preferred
Crenolanib is a potent and selective inhibitor of PDGFR / with Kd of 2 1 nM/3 2 nM in CHO cells also potently inhibits FLT3 sensitive to D842V mutation not V561D mutation 100-fold more selective for PDGFR than c-Kit VEGFR-2 TIE-2 FGFR-2 EGFR erbB2 and Src Crenolanib helps to induce mitophagy
Catalog No. 50-311-2232 Supplier Selleck Chemical LLC Supplier No. S273050MG
Crenolanib is a potent and selective inhibitor of PDGFR / with Kd of 2 1 nM/3 2 nM in CHO cells also potently inhibits FLT3 sensitive to D842V mutation not V561D mutation 100-fold more selective for PDGFR than c-Kit VEGFR-2 TIE-2 FGFR-2 EGFR erbB2 and Src Crenolanib helps to induce mitophagy