Learn More
MSC Crescent Adjustable Wrench, 18 in, 2-1/16 in Jaw Capacity, Alloy Steel, Chrome
Catalog No. 5000509543
Encompass
These adjustable wrenches are engineered from heat-treated forged alloy steel, ensuring toughness and durability. They feature a large knurl for easy adjustment and a hex jaw design to prevent slippage, securely gripping fasteners. The corrosion-resistant chrome finish prevents rusting, while the tapered handle offers improved grip and reduces fatigue, making them ideal for both professionals and DIY users.
  • Laser etched scale for SAE and metric identification.
  • Large knurl for tighter jaw fit and easier adjustment.
  • Hex jaw design reduces slippage.
  • Heat-treated forged alloy steel.
  • Tension spring stabilizes jaw.
  • Tapered handle for better grip and less weight.
  • Chrome plated finish resists rust and corrosion.
  • Large 2-1/16 in, 53 mm jaw capacity.
  • Reduces user fatigue.
  • Improved grip and leverage.
Catalog No. 50-005-09543 Supplier MSC Supplier No. 11931508
These adjustable wrenches are engineered from heat-treated forged alloy steel, ensuring toughness and durability. They feature a large knurl for easy adjustment and a hex jaw design to prevent slippage, securely gripping fasteners. The corrosion-resistant chrome finish prevents rusting, while the tapered handle offers improved grip and reduces fatigue, making them ideal for both professionals and DIY users.
  • Laser etched scale for SAE and metric identification.
  • Large knurl for tighter jaw fit and easier adjustment.
  • Hex jaw design reduces slippage.
  • Heat-treated forged alloy steel.
  • Tension spring stabilizes jaw.
  • Tapered handle for better grip and less weight.
  • Chrome plated finish resists rust and corrosion.
  • Large 2-1/16 in, 53 mm jaw capacity.
  • Reduces user fatigue.
  • Improved grip and leverage.