Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cresol mixture of isomers

Catalog No. 038691L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
03-869-1L 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03-869-1L Supplier Sigma Aldrich Supplier No. 038691L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3,3-dimethoxy-2-butanone is a biacetyl monoketal.3,3-dimethoxy-2-butanone may be used as starting reagent in the synthesis of 6-acety1-L,2-dihydro-2-oxo-3-pyridinecarboxylic acid
  • It may be used in the preparation of 4,4-dimethoxy-3-oxo-1-phenyl-1-pentene

Specifications

CAS 1319-77-3
MDL Number MFCD00151099
Quantity 25 g
Synonym Methylphenol tricresol
Percent Purity ∼85% (sum of isomers, GC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.