Learn More
Medchemexpress LLC Cresol red | 1733-12-6 | 99.3% | 100 MG
SDP
Catalog No. 5000411037
Encompass_Preferred
Cresol red, 97% is an anionic dye utilized as a pH indicator. It changes from orange to yellow at pH 1.8-2.0 and from yellow to purple at pH 7.0-8.8.
  • Anionic dye
  • Functions as a pH indicator
  • Changes color based on pH: orange to yellow at pH 1.8-2.0; yellow to purple at pH 7.0-8.8
  • For research use only; not sold to patients
Catalog No. 50-004-11037 Supplier Medchemexpress LLC Supplier No. HYD0015100MG
Cresol red, 97% is an anionic dye utilized as a pH indicator. It changes from orange to yellow at pH 1.8-2.0 and from yellow to purple at pH 7.0-8.8.
  • Anionic dye
  • Functions as a pH indicator
  • Changes color based on pH: orange to yellow at pH 1.8-2.0; yellow to purple at pH 7.0-8.8
  • For research use only; not sold to patients