Learn More
Bioworld Cresol Red, Free Acid, 5 g
Catalog No. 50199199
Encompass_Preferred
Loss on Drying(%)- <=5.0 Residue after Ignition(%)- <=0.1 Solubility (5mg/L water)(P/F)- Pass Visual Transition(P/F)- Pass
Catalog No. 50-199-199 Supplier Bioworld Supplier No. 217700111
Loss on Drying(%)- <=5.0 Residue after Ignition(%)- <=0.1 Solubility (5mg/L water)(P/F)- Pass Visual Transition(P/F)- Pass