Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cresol Red, Sigma-Aldrich

Catalog No. 01258939 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-939 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-939 Supplier MilliporeSigma Supelco Supplier No. 11447225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cresol Red, also known as o-cresol red, belongs to the phthalein and sulphonphthalein group of dyes. Cresol red changes color from orange to yellow at pH between 0. 2 to 1. 8 and yellow to violet at pH between 7 to 8. 8. In alkaline solutions, cresol red is present as a hydrophilic dianion. The dianion is small and is violet in color, possessing a larger conjugated system than the yellow hydroxyquinone.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1733-12-6
Molecular Weight (g/mol) 382.43 g/mol

Specifications

Color Brown/Red
Quantity 25 g
Grade Indicator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.